missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrophenylazo)resorcinol, 90%

Catalog No. AC151511000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151511000 100 g Glass bottle
AC151510250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151511000 Supplier Thermo Scientific Chemicals Supplier No. 151511000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 74-39-5 | C12H9N3O4 | 259.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74-39-5
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.22
MDL Number MFCD00007310
InChI Key BIFFKKLAAIGZBV-UHFFFAOYSA-N
Synonym p-nitrophenylazoresorcinol, p-diazoviolet, p-nitrobenzeneazoresorcinol, magneson i, magnezon i, 4-p-nitrophenylazo resorcinol, p-diazviolet, 2,4-dihydroxy-4'-nitroazobenzene, magneson, 4-4-nitrophenylazo resorcinol
PubChem CID 5717413
IUPAC Name (4E)-3-hydroxy-4-[(4-nitrophenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 185°C
Color Orange-Red to Red
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 16, 181
Merck Index 15, 5759
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in dilute aquous naoh
Formula Weight 259.21
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 4-(4-Nitrophenylazo)resorcinol, dye content ca 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 200-808-5
RTECSNumber VH2810000
TSCA TSCA

RUO – Research Use Only