missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrophenylazo)resorcinol, 90+%

Catalog No. AAA1223236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1223236 500 g
AAA1223214 25 g
AAA1223222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1223236 Supplier Thermo Scientific Chemicals Supplier No. A1223236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 74-39-5 | C12H9N3O4 | 259.22 g/mol

It is a derivitized resorcinol dye compound with an absorption maximum of 432 nm. Used as a pH indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a derivitized resorcinol dye compound with an absorption maximum of 432 nm. Used as a pH indicator.

Solubility
Soluble in dilute sodium hydroxide. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 74-39-5
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.22
MDL Number MFCD00007310
InChI Key BIFFKKLAAIGZBV-UHFFFAOYSA-N
Synonym p-nitrophenylazoresorcinol, p-diazoviolet, p-nitrobenzeneazoresorcinol, magneson i, magnezon i, 4-p-nitrophenylazo resorcinol, p-diazviolet, 2,4-dihydroxy-4'-nitroazobenzene, magneson, 4-4-nitrophenylazo resorcinol
PubChem CID 5717413
IUPAC Name (4E)-3-hydroxy-4-[(4-nitrophenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point ∼185°C (decomposition)
Quantity 500 g
Beilstein 674709
Merck Index 14,5695
Solubility Information Soluble in dilute sodium hydroxide. Insoluble in water.
Formula Weight 259.22
Percent Purity 95%
Chemical Name or Material 4- (4-Nitrophenylazo)resorcinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-808-5
RTECSNumber VH2810000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only