missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrophenylazo)phenol 97.0+%, TCI America™

Catalog No. N06535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06535G 5 g
N06531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06535G Supplier TCI America Supplier No. N06535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1435-60-5
Molecular Formula C12H9N3O3
Molecular Weight (g/mol) 243.22
MDL Number MFCD00161165
InChI Key ISHCVXKNSGUMPL-UHFFFAOYSA-N
PubChem CID 5355783
IUPAC Name 4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
SMILES [O-][N+](=O)C1=CC=C(NN=C2C=CC(=O)C=C2)C=C1

Specifications

Quantity 5 g
Synonym 4-Hydroxy-4′-nitroazobenzene
Formula Weight 243.22
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Nitrophenylazo)phenol