missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrophenylazo)orcinol 80.0+%, TCI America™

Catalog No. p-7133749
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03241G 1 g
N03245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03241G Supplier TCI America Supplier No. N03241G
Only null left

Chemical Identifiers

CAS 607-96-5
Molecular Formula C13H11N3O4
Molecular Weight (g/mol) 273.25
MDL Number MFCD00014710
InChI Key DJQKTCCAPLEDRJ-FYWRMAATSA-N
Synonym 4-nitrobenzene-azo-orcinol, 2,4-dihydroxy-6-methyl-4'-nitroazobenzene, 4-4-nitrophenylazo orcinol, 1,3-benzenediol, 5-methyl-4-2-4-nitrophenyl diazenyl, 4-4-nitrophenylazo-5-methylresorcinol, 5-methyl-4-4-nitrophenylazo resorcinol, 5-methyl-4-e-2-4-nitrophenyl diazen-1-yl benzene-1,3-diol, 1, 5-methyl-4-4-nitrophenyl azo, 2, 4-dihydroxy-6-methyl-4'-nitroazobenzene, 1,3-benzenediol, 5-methyl-4-4-nitrophenyl azo-
PubChem CID 6299952
IUPAC Name (4E)-3-hydroxy-5-methyl-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
SMILES CC1=CC(=O)C=C(O)\C1=N\NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 273.25
Percent Purity ≥80.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Nitrophenylazo)orcinol