missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrophenyl)morpholine 98.0+%, TCI America™

Catalog No. N11245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N11245G 5 g
N112425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N11245G Supplier TCI America Supplier No. N11245G
Only null left

Chemical Identifiers

CAS 10389-51-2
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
MDL Number MFCD00023317
InChI Key IAJDSUYFELYZCS-UHFFFAOYSA-N
Synonym 4-4-nitrophenyl morpholine, morpholine, 4-4-nitrophenyl, p-morpholinonitrobenzene, 4-p-nitrophenyl morpholine, 4-morpholinyl nitrobenzene, 4-morpholinonitrobenzene, n-p-nitrophenyl-morpholine, morpholine, 4-p-nitrophenyl, n-4-nitrophenyl morpholine
PubChem CID 25222
IUPAC Name 4-(4-nitrophenyl)morpholine
SMILES C1COCCN1C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 153°C
Quantity 5 g
Formula Weight 208.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Nitrophenyl)morpholine
EINECSNumber (5)-0871
RTECSNumber QE7405000
TSCA Yes