missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Morpholinylcarbonyl)benzeneboronic acid, 98%

Catalog No. p-7035409
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53126MD 250 mg
AAH5312603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53126MD Supplier Thermo Scientific Chemicals Supplier No. H53126MD
Only null left

CAS: 389621-84-5 | C11H14BNO4 | 235.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 389621-84-5
Molecular Formula C11H14BNO4
Molecular Weight (g/mol) 235.046
MDL Number MFCD03411952
InChI Key KMNLIQJXZPBCDU-UHFFFAOYSA-N
Synonym 4-morpholine-4-carbonyl phenylboronic acid, 4-morpholine-4-carbonyl phenyl boronic acid, 4-morpholinocarbonyl phenyl boronic acid, 4-4-boronobenzoyl morpholine, 4-morpholine-4-carbonyl benzeneboronic acid, boronic acid, 4-4-morpholinylcarbonyl phenyl, 4-boronophenyl morpholin-4-yl methanone, 4-morpholin-4-ylcarbonyl benzeneboronic acid, boronic acid,b-4-4-morpholinylcarbonyl phenyl
PubChem CID 2773546
IUPAC Name [4-(morpholine-4-carbonyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)N2CCOCC2)(O)O

Specifications

Melting Point 126°C to 132°C
Quantity 250 mg
Formula Weight 235.05
Percent Purity 98%
Chemical Name or Material 4-(4-Morpholinylcarbonyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only