missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Morpholinyl)pyridine, 97%

Catalog No. p-7052784
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5577906 5 g
AAH5577914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5577906 Supplier Thermo Scientific Chemicals Supplier No. H5577906
Only null left

CAS: 2767-91-1 | C9H12N2O | 164.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2767-91-1
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
MDL Number MFCD00704054
InChI Key QJWQYVJVCXMTJP-UHFFFAOYSA-N
Synonym 4-morpholinopyridine, 4-pyridin-4-yl morpholine, 4-4-pyridyl morpholine, morpholine, 4-4-pyridinyl, acmc-1cmri, 4-4-pyridinyl morpholine, 4-4-morpholinyl pyridine
PubChem CID 4145413
IUPAC Name 4-pyridin-4-ylmorpholine
SMILES C1COCCN1C2=CC=NC=C2

Specifications

Melting Point 102°C to 108°C
Color Yellow to Orange
Quantity 5 g
Formula Weight 164.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-(4-Morpholinyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only