missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Morpholinyl)benzeneboronic acid pinacol ester, 95%

Catalog No. p-7034257
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5192503 1 g
AAH5192506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5192503 Supplier Thermo Scientific Chemicals Supplier No. H5192503
Only null left

CAS: 568577-88-8 | C16H24BNO3 | 289.182 g/mol

4-(4-Morpholinyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Morpholinyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 568577-88-8
Molecular Formula C16H24BNO3
Molecular Weight (g/mol) 289.182
MDL Number MFCD04112544
InChI Key UCPALIMHMYIZPZ-UHFFFAOYSA-N
Synonym 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 4-morpholinophenylboronic acid pinacol ester, 4-morpholinophenylboronic acid, pinacol ester, 4-morpholin-4-yl-phenylboronic acid pinacol ester, 4-morpholin-4-ylbenzeneboronic acid pinacol ester, morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 4-morpholino phenylboronic acid pinacolate, 4-4-morpholinyl benzeneboronic acid pinacol ester
PubChem CID 2795361
IUPAC Name 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCOCC3

Specifications

Quantity 1 g
Formula Weight 289.18
Percent Purity 95%
Chemical Name or Material 4-(4-Morpholinyl)benzeneboronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only