missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazino)benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC18701DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC18701DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

86620-62-4
220.272
UCFZVQHKTRSZMM-UHFFFAOYSA-N
736532
CN1CCN(CC1)C2=CC=C(C=C2)C(=O)O
C12H16N2O2
MFCD02682063
4-4-methylpiperazin-1-yl benzoic acid, 4-4-methylpiperazino benzoic acid, 4-4-methyl-piperazin-1-yl-benzoic acid, 4-4-methylpiperazinyl benzoic acid, 4-4-methyl-piperazino benzoic acid, benzoic acid, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzoic acid, pubchem10481, ksc448o3p
4-(4-methylpiperazin-1-yl)benzoic acid

Specifications

4-(4-Methylpiperazino)benzoic acid
96.5
97%
C12H16N2O2
MFCD02682063
UCFZVQHKTRSZMM-UHFFFAOYSA-N
4-(4-methylpiperazin-1-yl)benzoic acid
736532
97%
86620-62-4
100.0
Amber Glass Bottle
1g
4-4-methylpiperazin-1-yl benzoic acid, 4-4-methylpiperazino benzoic acid, 4-4-methyl-piperazin-1-yl-benzoic acid, 4-4-methylpiperazinyl benzoic acid, 4-4-methyl-piperazino benzoic acid, benzoic acid, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzoic acid, pubchem10481, ksc448o3p
CN1CCN(CC1)C2=CC=C(C=C2)C(=O)O
220.272
220.27
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

Warning

SDS
Documents

Documents