missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Methylphenyl)-4-oxobutyric Acid 98.0+%, TCI America™
Supplier: TCI America M137725G
Specifications
| 4-(4-Methylphenyl)-4-oxobutyric Acid | |
| 128°C | |
| C11H12O3 | |
| 25 g | |
| OEEUWZITKKSXAZ-UHFFFAOYSA-N | |
| 4-(4-methylphenyl)-4-oxobutanoic acid | |
| 244162 | |
| ≥98.0% (GC,T) |
| 4619-20-9 | |
| White-Yellow | |
| MFCD00020541 | |
| 3-4-methylbenzoyl propionic acid, 4-oxo-4-p-tolyl butanoic acid, 4-4-methylphenyl-4-oxobutanoic acid, 4-4-methylphenyl-4-oxobutyric acid, 4-oxo-4-4-tolyl butyric acid, 4-oxo-4-p-tolylbutyric acid, 4-oxo-4-p-tolylbutanoic acid, 3-4-methylbenzoyl-propionic acid, acmc-1aooh, benzenebutanoic acid, 4-methyl-gamma-oxo | |
| CC1=CC=C(C=C1)C(=O)CCC(=O)O | |
| 192.214 | |
| 192.21 | |
| Crystalline Powder |
Chemical Identifiers
| 4619-20-9 | |
| 192.214 | |
| OEEUWZITKKSXAZ-UHFFFAOYSA-N | |
| 244162 | |
| CC1=CC=C(C=C1)C(=O)CCC(=O)O |
| C11H12O3 | |
| MFCD00020541 | |
| 3-4-methylbenzoyl propionic acid, 4-oxo-4-p-tolyl butanoic acid, 4-4-methylphenyl-4-oxobutanoic acid, 4-4-methylphenyl-4-oxobutyric acid, 4-oxo-4-4-tolyl butyric acid, 4-oxo-4-p-tolylbutyric acid, 4-oxo-4-p-tolylbutanoic acid, 3-4-methylbenzoyl-propionic acid, acmc-1aooh, benzenebutanoic acid, 4-methyl-gamma-oxo | |
| 4-(4-methylphenyl)-4-oxobutanoic acid |
Safety and Handling
EINECSNumber : (3)-4466
TSCA : No