missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylphenyl)-4-oxobutyric Acid 98.0+%, TCI America™

Catalog No. M13775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13775G 5 g
M137725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13775G Supplier TCI America Supplier No. M13775G

Chemical Identifiers

CAS 4619-20-9
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00020541
InChI Key OEEUWZITKKSXAZ-UHFFFAOYSA-N
Synonym 3-4-methylbenzoyl propionic acid, 4-oxo-4-p-tolyl butanoic acid, 4-4-methylphenyl-4-oxobutanoic acid, 4-4-methylphenyl-4-oxobutyric acid, 4-oxo-4-4-tolyl butyric acid, 4-oxo-4-p-tolylbutyric acid, 4-oxo-4-p-tolylbutanoic acid, 3-4-methylbenzoyl-propionic acid, acmc-1aooh, benzenebutanoic acid, 4-methyl-gamma-oxo
PubChem CID 244162
IUPAC Name 4-(4-methylphenyl)-4-oxobutanoic acid
SMILES CC1=CC=C(C=C1)C(=O)CCC(=O)O

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 5 g
Formula Weight 192.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Methylphenyl)-4-oxobutyric Acid
EINECSNumber (3)-4466
TSCA No