missing translation for 'onlineSavingsMsg'
Learn More

{4-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]phenyl}methanol, 97%, Thermo Scientific™

Catalog No. CC55809DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC55809DA 1g Amber Glass Bottle
CC55809DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC55809DA Supplier Thermo Scientific Chemicals Supplier No. CC55809DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 884507-50-0
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.343
MDL Number MFCD09064973
InChI Key YQUWAYPNHCWYNU-UHFFFAOYSA-N
Synonym 4-4-methylperhydro-1,4-diazepin-1-yl methyl phenyl methanol, 4-4-methyl-1,4-diazepan-1-yl methyl phenyl methanol, 4-4-methyl-1,4-diazaperhydroepinyl methyl phenyl methan-1-ol, benzenemethanol,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl methyl
PubChem CID 24229552
IUPAC Name [4-[(4-methyl-1,4-diazepan-1-yl)methyl]phenyl]methanol
SMILES CN1CCCN(CC1)CC2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 234.34
Percent Purity 97%
Chemical Name or Material {4-[(4-Methylperhydro-1, 4-diazepin-1-yl)methyl]phenyl}methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.