missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) 99.0+%, TCI America™

Catalog No. p-7132721
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
M1422500ML 500 mL
1 options

Catalog No. M1422500ML

Supplier: TCI America M1422500ML

Only null left

Chemical Identifiers

CAS 6864-37-5
Molecular Formula C15H30N2
Molecular Weight (g/mol) 238.42
MDL Number MFCD00075535
InChI Key IGSBHTZEJMPDSZ-UHFFFAOYNA-N
Synonym 4,4'-methylenebis 2-methylcyclohexanamine, bis 4-amino-3-methylcyclohexyl methane, cyclohexanamine, 4,4'-methylenebis 2-methyl, 4,4'-methylenebis 2-methylcyclohexylamine, 3,3'-dimethyl-4,4'-diaminodicyclohexylmethane, 2,2'-dimethyl-4,4'-methylenebis cyclohexylamine, 4-4-amino-3-methylcyclohexyl methyl-2-methylcyclohexan-1-amine, 4,4'-methylenebis 2-methylcyclohexyl-amine, 2,2'-dimethyl-4,4'methylenebis cyclohexylamine
PubChem CID 91555
IUPAC Name 4-[(4-amino-3-methylcyclohexyl)methyl]-2-methylcyclohexan-1-amine
SMILES CC1CC(CC2CCC(N)C(C)C2)CCC1N

Specifications

Color Yellow
Quantity 500 mL
UN Number 2927
Formula Weight 238.42
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4′-Methylenebis(2-methylcyclohexylamine) (mixture of isomers)
EINECSNumber (4)-0102
TSCA Yes