missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) 99.0+%, TCI America™

Catalog No. M142225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M142225ML 25 mL
M1422500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M142225ML Supplier TCI America Supplier No. M142225ML
Only null left

Chemical Identifiers

CAS 6864-37-5
Molecular Formula C15H30N2
Molecular Weight (g/mol) 238.42
MDL Number MFCD00075535
InChI Key IGSBHTZEJMPDSZ-UHFFFAOYNA-N
Synonym 4,4'-methylenebis 2-methylcyclohexanamine, bis 4-amino-3-methylcyclohexyl methane, cyclohexanamine, 4,4'-methylenebis 2-methyl, 4,4'-methylenebis 2-methylcyclohexylamine, 3,3'-dimethyl-4,4'-diaminodicyclohexylmethane, 2,2'-dimethyl-4,4'-methylenebis cyclohexylamine, 4-4-amino-3-methylcyclohexyl methyl-2-methylcyclohexan-1-amine, 4,4'-methylenebis 2-methylcyclohexyl-amine, 2,2'-dimethyl-4,4'methylenebis cyclohexylamine
PubChem CID 91555
IUPAC Name 4-[(4-amino-3-methylcyclohexyl)methyl]-2-methylcyclohexan-1-amine
SMILES CC1CC(CC2CCC(N)C(C)C2)CCC1N

Specifications

Color Yellow
Quantity 25 mL
UN Number 2927
Formula Weight 238.42
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4′-Methylenebis(2-methylcyclohexylamine) (mixture of isomers)
EINECSNumber (4)-0102
TSCA Yes