missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2-methylcyclohexylamine), 99%, mixture of isomers, Thermo Scientific™

Catalog No. p-4813039
Change view
Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC450242500 250mL Glass bottle
AC450240010 1L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450242500 Supplier Thermo Scientific Chemicals Supplier No. 450242500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 6864-37-5
Molecular Formula C15H30N2
Molecular Weight (g/mol) 238.42
MDL Number MFCD00075535
InChI Key IGSBHTZEJMPDSZ-UHFFFAOYNA-N
Synonym 4,4'-methylenebis 2-methylcyclohexanamine, bis 4-amino-3-methylcyclohexyl methane, cyclohexanamine, 4,4'-methylenebis 2-methyl, 4,4'-methylenebis 2-methylcyclohexylamine, 3,3'-dimethyl-4,4'-diaminodicyclohexylmethane, 2,2'-dimethyl-4,4'-methylenebis cyclohexylamine, 4-4-amino-3-methylcyclohexyl methyl-2-methylcyclohexan-1-amine, 4,4'-methylenebis 2-methylcyclohexyl-amine, 2,2'-dimethyl-4,4'methylenebis cyclohexylamine
PubChem CID 91555
IUPAC Name 4-[(4-amino-3-methylcyclohexyl)methyl]-2-methylcyclohexan-1-amine
SMILES CC1CC(CC2CCC(N)C(C)C2)CCC1N

Specifications

Density 0.9400g/mL
Boiling Point 93.0°C to 100.0°C
Flash Point 113°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 250mL
Specific Gravity 0.94
Viscosity 142 mPa.s (20°C)
Formula Weight 238.41
Percent Purity 99%
Chemical Name or Material 4, 4'-Methylenebis(2-methylcyclohexylamine)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
EINECSNumber 229-962-1

RUO – Research Use Only