missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methyl-1-piperazinyl)aniline 98.0+%, TCI America™

Catalog No. M21545G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21545G 5 g
M21541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21545G Supplier TCI America Supplier No. M21545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16153-81-4
Molecular Formula C11H18N3
Molecular Weight (g/mol) 192.29
MDL Number MFCD00172703
InChI Key MOZNZNKHRXRLLF-UHFFFAOYSA-O
Synonym 4-4-methylpiperazino aniline, 4-4-methylpiperazin-1-yl aniline, 4-methyl-1-4-aminophenyl piperazine, 4-4-methyl-1-piperazinyl aniline, 4-4-methylpiperazin-1-yl phenylamine, 1-4-aminophenyl-4-methylpiperazine, 4-4-methyl-piperazin-1-yl-phenylamine, 4-4-methylpiperazin-1-yl benzenamine, benzenamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenylamine
PubChem CID 737253
IUPAC Name 4-(4-aminophenyl)-1-methylpiperazin-1-ium
SMILES C[NH+]1CCN(CC1)C1=CC=C(N)C=C1

Specifications

Melting Point 91°C
Boiling Point 180°C
Quantity 5 g
UN Number 3263
Formula Weight 191.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Methyl-1-piperazinyl)aniline