missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methyl-1-piperazinyl)aniline, 97%

Catalog No. AAH2668406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2668406 5 g
AAH2668403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2668406 Supplier Thermo Scientific Chemicals Supplier No. H2668406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16153-81-4 | C11H18N3 | 192.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16153-81-4
Molecular Formula C11H18N3
Molecular Weight (g/mol) 192.29
MDL Number MFCD00172703
InChI Key MOZNZNKHRXRLLF-UHFFFAOYSA-O
Synonym 4-4-methylpiperazino aniline, 4-4-methylpiperazin-1-yl aniline, 4-methyl-1-4-aminophenyl piperazine, 4-4-methyl-1-piperazinyl aniline, 4-4-methylpiperazin-1-yl phenylamine, 1-4-aminophenyl-4-methylpiperazine, 4-4-methyl-piperazin-1-yl-phenylamine, 4-4-methylpiperazin-1-yl benzenamine, benzenamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenylamine
PubChem CID 737253
IUPAC Name 4-(4-methylpiperazin-1-yl)aniline
SMILES C[NH+]1CCN(CC1)C1=CC=C(N)C=C1

Specifications

Melting Point 89°C to 94°C
Quantity 5 g
UN Number UN3263
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material 4-(4-Methyl-1-piperazinyl)aniline
Hazard Category H314
Hazard Statement GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only