missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine, 98%

CAS: 13104-56-8 | C22H17N3O | 339.398 g/mol

Supplier:  Thermo Scientific Chemicals B2548602

 View more versions of this product

Catalog No. AAB2548602


Only null left
Explore available promotions

Description

Description

4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-(4-Methoxyphenyl)-2,2′:6′,2′′-terpyridine is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine
161°C to 163°C
MFCD06796987
4'-4-methoxyphenyl-2,2':6',2-terpyridine, 4-4-methoxyphenyl-2,6-dipyridin-2-ylpyridine, 4-4-methoxyphenyl-2,6-bis pyridin-2-yl pyridine, 4-p-methoxyphenyl-2,6-di 2-pyridyl pyridine, qaewaywlmregra-uhfffaoysa, 4'-4-methoxyphenyl-2,2',6',2-terpyridine, 4'-4-methoxyphenyl-2,2':6',2'-terpyridine, 4-4-methoxyphenyl-2,2:6,2-terpyridine, 4-4-methoxyphenyl-2,6-di pyridin-2-yl pyridine, 1-2,6-di 2-pyridyl 4-pyridyl-4-methoxybenzene
QAEWAYWLMREGRA-UHFFFAOYSA-N
4-(4-methoxyphenyl)-2,6-dipyridin-2-ylpyridine
630929
98%
13104-56-8
C22H17N3O
0.5 g
Insoluble in water.
COC1=CC=C(C=C1)C2=CC(=NC(=C2)C3=CC=CC=N3)C4=CC=CC=N4
339.398
339.4

Chemical Identifiers

13104-56-8
339.398
QAEWAYWLMREGRA-UHFFFAOYSA-N
630929
COC1=CC=C(C=C1)C2=CC(=NC(=C2)C3=CC=CC=N3)C4=CC=CC=N4
C22H17N3O
MFCD06796987
4'-4-methoxyphenyl-2,2':6',2-terpyridine, 4-4-methoxyphenyl-2,6-dipyridin-2-ylpyridine, 4-4-methoxyphenyl-2,6-bis pyridin-2-yl pyridine, 4-p-methoxyphenyl-2,6-di 2-pyridyl pyridine, qaewaywlmregra-uhfffaoysa, 4'-4-methoxyphenyl-2,2',6',2-terpyridine, 4'-4-methoxyphenyl-2,2':6',2'-terpyridine, 4-4-methoxyphenyl-2,2:6,2-terpyridine, 4-4-methoxyphenyl-2,6-di pyridin-2-yl pyridine, 1-2,6-di 2-pyridyl 4-pyridyl-4-methoxybenzene
4-(4-methoxyphenyl)-2,6-dipyridin-2-ylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only