missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine, 98%

Catalog No. AAB2548677
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2548677 0.1 g
AAB2548602 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2548677 Supplier Thermo Scientific Chemicals Supplier No. B2548677
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13104-56-8 | C22H17N3O | 339.398 g/mol

4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-(4-Methoxyphenyl)-2,2′:6′,2′′-terpyridine is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13104-56-8
Molecular Formula C22H17N3O
Molecular Weight (g/mol) 339.398
MDL Number MFCD06796987
InChI Key QAEWAYWLMREGRA-UHFFFAOYSA-N
Synonym 4'-4-methoxyphenyl-2,2':6',2-terpyridine, 4-4-methoxyphenyl-2,6-dipyridin-2-ylpyridine, 4-4-methoxyphenyl-2,6-bis pyridin-2-yl pyridine, 4-p-methoxyphenyl-2,6-di 2-pyridyl pyridine, qaewaywlmregra-uhfffaoysa, 4'-4-methoxyphenyl-2,2',6',2-terpyridine, 4'-4-methoxyphenyl-2,2':6',2'-terpyridine, 4-4-methoxyphenyl-2,2:6,2-terpyridine, 4-4-methoxyphenyl-2,6-di pyridin-2-yl pyridine, 1-2,6-di 2-pyridyl 4-pyridyl-4-methoxybenzene
PubChem CID 630929
IUPAC Name 4-(4-methoxyphenyl)-2,6-dipyridin-2-ylpyridine
SMILES COC1=CC=C(C=C1)C2=CC(=NC(=C2)C3=CC=CC=N3)C4=CC=CC=N4

Specifications

Melting Point 161°C to 163°C
Quantity 0.1 g
Solubility Information Insoluble in water.
Formula Weight 339.4
Percent Purity 98%
Chemical Name or Material 4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only