missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Formylphenyl)morpholine 98.0+%, TCI America™

Catalog No. M16251G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16251G 1 g
M16255G 5 g
M162525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M16251G Supplier TCI America Supplier No. M16251G
Only null left

Chemical Identifiers

CAS 1204-86-0
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00735826
InChI Key FOAQOAXQMISINY-UHFFFAOYSA-N
Synonym 4-morpholinobenzaldehyde, 4-morpholin-4-yl-benzaldehyde, 4-4-morpholinyl benzaldehyde, 4-4-formylphenyl morpholine, 4-morpholin-4-yl benzaldehyde, 4-morpholinobenzenecarbaldehyde, benzaldehyde, 4-4-morpholinyl, akos bc-2976, acmc-1bqjc, 4-morpholinylbenzaldehyde
PubChem CID 291349
IUPAC Name 4-(morpholin-4-yl)benzaldehyde
SMILES O=CC1=CC=C(C=C1)N1CCOCC1

Specifications

Melting Point 63°C
Quantity 1 g
Formula Weight 191.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Formylphenyl)morpholine