missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)imidazole, 97%

Catalog No. AAH3150306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3150306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3150306 Supplier Thermo Scientific Chemicals Supplier No. H3150306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65020-70-4 | C9H7FN2 | 162.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65020-70-4
Molecular Formula C9H7FN2
Molecular Weight (g/mol) 162.167
MDL Number MFCD00297041
InChI Key QYDAFWKRBZYBOB-UHFFFAOYSA-N
Synonym 4-4-fluorophenyl-1h-imidazole, 5-4-fluorophenyl-1h-imidazole, 1h-imidazole,5-4-fluorophenyl, 4-4-fluoro-phenyl-1h-imidazole, regid_for_cid_657876, 5-4-fluorophenyl-3h-imidazole, 4-4-fluorophenyl-1h-imidazole, 9, 1h-imidazole, 5-4-fluorophenyl, 3sq
PubChem CID 657876
IUPAC Name 5-(4-fluorophenyl)-1H-imidazole
SMILES C1=CC(=CC=C1C2=CN=CN2)F

Specifications

Melting Point 127°C to 131°C
Quantity 5 g
Formula Weight 162.17
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorophenyl)imidazole
Hazard Category H302-H318
Hazard Statement GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only