missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde 98.0+%, TCI America™
$52.19 - $158.44
Chemical Identifiers
| CAS | 147118-37-4 |
|---|---|
| Molecular Formula | C16H18FN3O3S |
| Molecular Weight (g/mol) | 351.396 |
| MDL Number | MFCD08458342 |
| InChI Key | WOCOTUDOVSLFOB-UHFFFAOYSA-N |
| Synonym | n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde |
| PubChem CID | 10473133 |
| IUPAC Name | N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide |
| SMILES | CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C |
Chemical Identifiers
| 147118-37-4 | |
| 351.396 | |
| WOCOTUDOVSLFOB-UHFFFAOYSA-N | |
| 10473133 | |
| CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C |
| C16H18FN3O3S | |
| MFCD08458342 | |
| n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde | |
| N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide |
Specifications
| 147118-37-4 | |
| White | |
| MFCD08458342 | |
| n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde | |
| CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C | |
| 351.396 | |
| 351.40 | |
| Crystalline Powder |
| 180°C | |
| C16H18FN3O3S | |
| 5 g | |
| WOCOTUDOVSLFOB-UHFFFAOYSA-N | |
| N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide | |
| 10473133 | |
| ≥98.0% (HPLC,N) | |
| 4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde |
Safety and Handling
TSCA : No