missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde 98.0+%, TCI America™

Catalog No. F08675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08675G 5 g
F086725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08675G Supplier TCI America Supplier No. F08675G

Chemical Identifiers

CAS 147118-37-4
Molecular Formula C16H18FN3O3S
Molecular Weight (g/mol) 351.396
MDL Number MFCD08458342
InChI Key WOCOTUDOVSLFOB-UHFFFAOYSA-N
Synonym n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde
PubChem CID 10473133
IUPAC Name N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
SMILES CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Formula Weight 351.40
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde