missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)-4-hydroxypiperidine, 97%

Catalog No. AAH6389203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6389203 1 g
AAH6389206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6389203 Supplier Thermo Scientific Chemicals Supplier No. H6389203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3888-65-1 | C11H14FNO | 195.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3888-65-1
Molecular Formula C11H14FNO
Molecular Weight (g/mol) 195.24
MDL Number MFCD00023146
InChI Key QXWRXWPNHLIZBV-UHFFFAOYSA-N
PubChem CID 77497
IUPAC Name 4-(4-fluorophenyl)piperidin-4-ol
SMILES OC1(CCNCC1)C1=CC=C(F)C=C1

Specifications

Quantity 1 g
Synonym 4-4-fluorophenyl piperidin-4-ol, 4-piperidinol, 4-4-fluorophenyl, 4-4-fluorophenyl-4-hydroxypiperidine, 4-4-fluorophenyl-4-piperidinol, 4-fluorophenyl-4-hydroxylpiperidinehydrochloride, 4-4-fluoro-phenyl piperidin-4-ol, 4-piperidinol,4-4-fluorophenyl, 4-4-fluorophenyl-4hydroxypiperidine, 4-hydroxy-4-4-fluorophenyl piperidine, 4-4-fluorophenyl-4-hydroxy piperidine
Formula Weight 195.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Fluorophenyl)-4-hydroxypiperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-431-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only