missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorobenzyloxy)benzaldehyde, 97%

Catalog No. AAA1765706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1765706 5 g
AAA1765714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1765706 Supplier Thermo Scientific Chemicals Supplier No. A1765706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56442-17-2 | C14H11FO2 | 230.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56442-17-2
Molecular Formula C14H11FO2
Molecular Weight (g/mol) 230.24
MDL Number MFCD00173650
InChI Key QGULWBQOCMQNFD-UHFFFAOYSA-N
Synonym 4-4-fluorobenzyloxy benzaldehyde, 4-4-fluorobenzyl oxy benzaldehyde, 4-4-fluorophenyl methoxy benzaldehyde, acmc-1b0gw, cbmicro_008348, 4-4-fluorobenzyl-oxy benzaldehyde, 4-4-fluoro-benzyloxy-benzaldehyde, benzaldehyde,4-4-fluorophenyl methoxy
PubChem CID 735860
IUPAC Name 4-[(4-fluorophenyl)methoxy]benzaldehyde
SMILES FC1=CC=C(COC2=CC=C(C=O)C=C2)C=C1

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 230.24
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorobenzyloxy)benzaldehyde
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only