missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%

Catalog No. AAB2314203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2314203 1 g
AAB2314206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2314203 Supplier Thermo Scientific Chemicals Supplier No. B2314203

CAS: 25519-78-2 | C12H15ClFNO | 243.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25519-78-2
Molecular Formula C12H15ClFNO
Molecular Weight (g/mol) 243.71
MDL Number MFCD00044912
InChI Key GPKDBZQZPNOBGM-UHFFFAOYSA-N
Synonym 4-4-fluorobenzoyl piperidine hydrochloride, 4-fluorophenyl piperidin-4-yl methanone hydrochloride, 4-fluorophenyl 4-piperidinyl ketone hydrochloride, p-fluorophenyl 4-piperidyl ketone hydrochloride, 4-fluorophenyl 4-piperidyl methanone hydrochloride, 4-fluoro-phenyl-piperidin-4-yl-methanone hydrochloride, 4-4-fluorobenzoyl piperidine hcl, 4-p-fluorobenzoyl piperidine hydrochloride, 4-4'-fluorobenzoyl piperidine hydrochloride, methanone, 4-fluorophenyl-4-piperidinyl-, hydrochloride
PubChem CID 3084438
IUPAC Name (4-fluorophenyl)-piperidin-4-ylmethanone;hydrochloride
SMILES [H+].[Cl-].FC1=CC=C(C=C1)C(=O)C1CCNCC1

Specifications

Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 243.71
Percent Purity 98%
Chemical Name or Material 4-(4-Fluorobenzoyl)piperidine hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 247-070-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only