missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorobenzoyl)butyric Acid 98.0+%, TCI America™

Catalog No. F04761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04761G 1 g
F04765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04761G Supplier TCI America Supplier No. F04761G

Chemical Identifiers

CAS 149437-76-3
Molecular Formula C11H11FO3
Molecular Weight (g/mol) 210.204
MDL Number MFCD00667222
InChI Key ZBQROUOOMAMCQW-UHFFFAOYSA-N
Synonym 4-4-fluorobenzoyl butyric acid, 5-4-fluorophenyl-5-oxopentanoic acid, 4-4-fluorobenzoyl butyricacid, 5-4-fluorophenyl-5-oxovaleric acid, 4-fluorobenzoylbutyric acid, 5-4'-fluorophenyl-5-oxopentanoic acid, 4-4-fluorobenzoyl butanoic acid, 4-4-fluorobenzoyl-butyric acid, 4-fluorophenyl-5-oxopentanoic acid, 4-4'-fluorobenzoyl butyric acid
PubChem CID 689096
IUPAC Name 5-(4-fluorophenyl)-5-oxopentanoic acid
SMILES C1=CC(=CC=C1C(=O)CCCC(=O)O)F

Specifications

Melting Point 143°C
Color White
Quantity 1 g
Formula Weight 210.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Fluorobenzoyl)butyric Acid