missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dipyridyl, 98%

Catalog No. p-4510397
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC276200050 5 g Glass bottle
AC276200250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC276200050 Supplier Thermo Scientific Chemicals Supplier No. 276200050
Only null left

CAS: 553-26-4 | C10H8N2 | 156.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 553-26-4
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006416
InChI Key MWVTWFVJZLCBMC-UHFFFAOYSA-N
Synonym 4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
PubChem CID 11107
ChEBI CHEBI:30985
IUPAC Name 4-pyridin-4-ylpyridine
SMILES C1=CC(=CC=N1)C1=CC=NC=C1

Specifications

Melting Point 110.0°C to 114.0°C
Color White to Beige
Boiling Point 305.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C5H4N)-(C5H4N)
Quantity 5 g
Beilstein 23,2
Fieser 16,25
Merck Index 15,3389
Solubility Information Solubility in water: 4,5g/L (25°C). Other solubilities: freely soluble in alcohol, benzene, chloroform,, ether,
Formula Weight 156.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Dipyridyl
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist
EINECSNumber 209-036-3

RUO – Research Use Only