missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethylcyclohexanone 98.0+%, TCI America™

Catalog No. p-7128125
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41491G 1 g
D41495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41491G Supplier TCI America Supplier No. D41491G
Only null left

Chemical Identifiers

CAS 4255-62-3
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00234965
InChI Key PXQMSTLNSHMSJB-UHFFFAOYSA-N
Synonym 4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanone
PubChem CID 138166
IUPAC Name 4,4-dimethylcyclohexan-1-one
SMILES CC1(CCC(=O)CC1)C

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 73°C
Quantity 1 g
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Dimethylcyclohexanone