missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethylcyclohexanone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 44°C |
| Color | White-Yellow |
| Boiling Point | 73°C |
| Quantity | 1 g |
| Formula Weight | 126.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4-Dimethylcyclohexanone |
Safety and Handling
| TSCA | No |