missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethylbiphenyl 97.0+%, TCI America™
Supplier: TCI America D12345G
Specifications
| 4,4′-Dimethylbiphenyl | |
| 122°C | |
| C14H14 | |
| 5 g | |
| RZTDESRVPFKCBH-UHFFFAOYSA-N | |
| 1-methyl-4-(4-methylphenyl)benzene | |
| 11941 | |
| ≥97.0% (GC) |
| 613-33-2 | |
| White-Yellow | |
| MFCD00008545 | |
| 4,4'-dimethylbiphenyl, 4,4'-bitolyl, 4,4'-dimethyldiphenyl, bi-p-tolyl, di-p-tolyl, p,p'-bitolyl, 4,4'-dimethyl-1,1'-biphenyl, 4,4'-ditolyl, 1,1'-biphenyl, 4,4'-dimethyl, 4,4'-dimethyl-biphenyl | |
| CC1=CC=C(C=C1)C2=CC=C(C=C2)C | |
| 182.266 | |
| 182.27 | |
| Crystalline Powder |
Chemical Identifiers
| 613-33-2 | |
| 182.266 | |
| RZTDESRVPFKCBH-UHFFFAOYSA-N | |
| 11941 | |
| CC1=CC=C(C=C1)C2=CC=C(C=C2)C |
| C14H14 | |
| MFCD00008545 | |
| 4,4'-dimethylbiphenyl, 4,4'-bitolyl, 4,4'-dimethyldiphenyl, bi-p-tolyl, di-p-tolyl, p,p'-bitolyl, 4,4'-dimethyl-1,1'-biphenyl, 4,4'-ditolyl, 1,1'-biphenyl, 4,4'-dimethyl, 4,4'-dimethyl-biphenyl | |
| 1-methyl-4-(4-methylphenyl)benzene |
Safety and Handling
TSCA : No