missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbiphenyl 97.0+%, TCI America™

Catalog No. D12341G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12341G 1 g
D12345G 5 g
D123425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D12341G Supplier TCI America Supplier No. D12341G

Chemical Identifiers

CAS 613-33-2
Molecular Formula C14H14
Molecular Weight (g/mol) 182.266
MDL Number MFCD00008545
InChI Key RZTDESRVPFKCBH-UHFFFAOYSA-N
Synonym 4,4'-dimethylbiphenyl, 4,4'-bitolyl, 4,4'-dimethyldiphenyl, bi-p-tolyl, di-p-tolyl, p,p'-bitolyl, 4,4'-dimethyl-1,1'-biphenyl, 4,4'-ditolyl, 1,1'-biphenyl, 4,4'-dimethyl, 4,4'-dimethyl-biphenyl
PubChem CID 11941
IUPAC Name 1-methyl-4-(4-methylphenyl)benzene
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 1 g
Formula Weight 182.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethylbiphenyl