missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbiphenyl, 99%

Catalog No. AAB2508903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2508903 1 g
AAB2508906 5 g
AAB2508914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2508903 Supplier Thermo Scientific Chemicals Supplier No. B2508903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 613-33-2 | C14H14 | 182.266 g/mol

4,4'-Dimethylbiphenyl is used in the preparation of biphenyl-4,4’-dicarboxylic acid, which acts as a monomer utilized in the synthesis of linear long chain polymer through polycondensation reaction. It is used to prepare 4-(4'-methylphenyl)benzaldehyde by selective photoxygenation reaction using 9-phenyl-10-methylacridinium perchlorate as a catalyst under visible light irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethylbiphenyl is used in the preparation of biphenyl-4,4’-dicarboxylic acid, which acts as a monomer utilized in the synthesis of linear long chain polymer through polycondensation reaction. It is used to prepare 4-(4′-methylphenyl)benzaldehyde by selective photoxygenation reaction using 9-phenyl-10-methylacridinium perchlorate as a catalyst under visible light irradiation.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 613-33-2
Molecular Formula C14H14
Molecular Weight (g/mol) 182.266
MDL Number MFCD00008545
InChI Key RZTDESRVPFKCBH-UHFFFAOYSA-N
Synonym 4,4'-dimethylbiphenyl, 4,4'-bitolyl, 4,4'-dimethyldiphenyl, bi-p-tolyl, di-p-tolyl, p,p'-bitolyl, 4,4'-dimethyl-1,1'-biphenyl, 4,4'-ditolyl, 1,1'-biphenyl, 4,4'-dimethyl, 4,4'-dimethyl-biphenyl
PubChem CID 11941
IUPAC Name 1-methyl-4-(4-methylphenyl)benzene
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C

Specifications

Melting Point 118°C to 122°C
Boiling Point 295°C
Quantity 1 g
Beilstein 1856053
Solubility Information Insoluble in water.
Formula Weight 182.27
Percent Purity 99%
Chemical Name or Material 4,4'-Dimethylbiphenyl
EINECSNumber 210-337-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only