missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzil 99.0+%, TCI America™

Catalog No. D187525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D187525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D187525G Supplier TCI America Supplier No. D187525G

Chemical Identifiers

CAS 3457-48-5
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00008554
InChI Key BCWCEHMHCDCJAD-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzil, 1,2-di-p-tolylethane-1,2-dione, p-tolil, 1,2-bis 4-methylphenyl ethane-1,2-dione, ethanedione, bis 4-methylphenyl, ethandione, bis p-tolyl, bis 4-methylphenyl ethane-1,2-dione, 1,2-bis 4-methylphenyl-1,2-ethanedione, di-p-tolylethanedione, 4,4/'-dimethylbenzil
PubChem CID 76996
IUPAC Name 1,2-bis(4-methylphenyl)ethane-1,2-dione
SMILES CC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 105°C
Boiling Point 211°C
Quantity 25 g
Formula Weight 238.29
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethylbenzil