missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzil, 98%

Catalog No. p-7024242
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1857603 1 g
AAA1857606 5 g
AAA1857614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1857603 Supplier Thermo Scientific Chemicals Supplier No. A1857603
Only null left

CAS: 3457-48-5 | C16H14O2 | 238.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3457-48-5
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00008554
InChI Key BCWCEHMHCDCJAD-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzil, 1,2-di-p-tolylethane-1,2-dione, p-tolil, 1,2-bis 4-methylphenyl ethane-1,2-dione, ethanedione, bis 4-methylphenyl, ethandione, bis p-tolyl, bis 4-methylphenyl ethane-1,2-dione, 1,2-bis 4-methylphenyl-1,2-ethanedione, di-p-tolylethanedione, 4,4/'-dimethylbenzil
PubChem CID 76996
IUPAC Name 1,2-bis(4-methylphenyl)ethane-1,2-dione
SMILES CC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 102°C to 104°C
Quantity 1 g
Formula Weight 238.29
Percent Purity 98%
Chemical Name or Material 4,4'-Dimethylbenzil
EINECSNumber 222-388-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only