missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, 97%

Catalog No. AC408110025
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408110025 2.5 g Glass bottle
AC408110100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC408110025 Supplier Thermo Scientific Chemicals Supplier No. 408110025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 845-46-5 | C15H14N3NaO2 | 291.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 845-46-5
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.27
MDL Number MFCD00020350
InChI Key OSCKRHPYZNTEIO-UHFFFAOYSA-M
Synonym p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid
PubChem CID 23674498
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Quantity 2.5 g
Formula Weight 291.27
Percent Purity 98%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, 98%
Show More Show Less

RUO – Research Use Only