missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethyl-trans-stilbene 99.0+%, TCI America™
$398.90 - $398.90
Chemical Identifiers
| CAS | 18869-29-9 |
|---|---|
| Molecular Formula | C16H16 |
| Molecular Weight (g/mol) | 208.304 |
| MDL Number | MFCD00136904 |
| InChI Key | KINZBJFIDFZQCB-VAWYXSNFSA-N |
| PubChem CID | 5462964 |
| IUPAC Name | 1-methyl-4-[(E)-2-(4-methylphenyl)ethenyl]benzene |
| SMILES | CC1=CC=C(C=C1)C=CC2=CC=C(C=C2)C |
Chemical Identifiers
| 18869-29-9 | |
| 208.304 | |
| KINZBJFIDFZQCB-VAWYXSNFSA-N | |
| 1-methyl-4-[(E)-2-(4-methylphenyl)ethenyl]benzene |
| C16H16 | |
| MFCD00136904 | |
| 5462964 | |
| CC1=CC=C(C=C1)C=CC2=CC=C(C=C2)C |
Specifications
| 18869-29-9 | |
| White | |
| MFCD00136904 | |
| KINZBJFIDFZQCB-VAWYXSNFSA-N | |
| 1-methyl-4-[(E)-2-(4-methylphenyl)ethenyl]benzene | |
| 5462964 | |
| ≥99.0% (GC) | |
| 4,4′-Dimethyl-trans-stilbene |
| 183°C | |
| C16H16 | |
| 5 g | |
| CC1=CC=C(C=C1)C=CC2=CC=C(C=C2)C | |
| 208.304 | |
| 208.30 | |
| Crystalline Powder |
Safety and Handling
TSCA : No