missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-2-pentanone, 99%
CAS: 590-50-1 | C7H14O | 114.19 g/mol
$284.22 - $284.22
Chemical Identifiers
| CAS | 590-50-1 |
|---|---|
| Molecular Formula | C7H14O |
| Molecular Weight (g/mol) | 114.19 |
| InChI Key | AZASWMGVGQEVCS-UHFFFAOYSA-N |
| PubChem CID | 11546 |
| IUPAC Name | 4,4-dimethylpentan-2-one |
| SMILES | CC(=O)CC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC159710050
|
Thermo Scientific Chemicals
159710050 |
5 g |
Each for $284.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 590-50-1 | |
| 114.19 | |
| 11546 | |
| CC(=O)CC(C)(C)C |
| C7H14O | |
| AZASWMGVGQEVCS-UHFFFAOYSA-N | |
| 4,4-dimethylpentan-2-one |
Specifications
| 590-50-1 | |
| 100.0 | |
| 0.8040g/mL | |
| 18°C | |
| 99% | |
| C7H14O | |
| (CH3)3CCH2COCH3 | |
| 01,702 | |
| 4,4-dimethyl-2-pentanone, 2-pentanone, 4,4-dimethyl, methyl neopentyl ketone, neopentyl methyl ketone, methyl neoamyl ketone, 4,4-dimethyl-pentan-2-one, 2-pentanone,4,4-dimethyl, acmc-1awgm, neo-c5h11coch3 | |
| CC(=O)CC(C)(C)C | |
| 114.19 | |
| 114.19 | |
| Liquid |
| 98.5 | |
| Colorless to Yellow | |
| 125.0°C to 130.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4025 to 1.4045 | |
| 5 g | |
| 0.804 | |
| AZASWMGVGQEVCS-UHFFFAOYSA-N | |
| 4,4-dimethylpentan-2-one | |
| 11546 | |
| 99% | |
| 4, 4-Dimethyl-2-pentanone |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Danger
EINECSNumber : 209-685-2
RUO – Research Use Only