missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-pentanone, 99%

Catalog No. AC159710050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC159710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC159710050 Supplier Thermo Scientific Chemicals Supplier No. 159710050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 590-50-1 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 590-50-1
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
InChI Key AZASWMGVGQEVCS-UHFFFAOYSA-N
PubChem CID 11546
IUPAC Name 4,4-dimethylpentan-2-one
SMILES CC(=O)CC(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8040g/mL
Boiling Point 125.0°C to 130.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4025 to 1.4045
Linear Formula (CH3)3CCH2COCH3
Quantity 5 g
Beilstein 01,702
Specific Gravity 0.804
Synonym 4,4-dimethyl-2-pentanone, 2-pentanone, 4,4-dimethyl, methyl neopentyl ketone, neopentyl methyl ketone, methyl neoamyl ketone, 4,4-dimethyl-pentan-2-one, 2-pentanone,4,4-dimethyl, acmc-1awgm, neo-c5h11coch3
Formula Weight 114.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4, 4-Dimethyl-2-pentanone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 209-685-2

RUO – Research Use Only