missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-oxazoline 97.0+%, TCI America™

Catalog No. D27425ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27425ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27425ML Supplier TCI America Supplier No. D27425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30093-99-3
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00014507
InChI Key KOAMXHRRVFDWRQ-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl
PubChem CID 121630
IUPAC Name 4,4-dimethyl-4,5-dihydro-1,3-oxazole
SMILES CC1(C)COC=N1

Specifications

Color Colorless
Boiling Point 100°C
Quantity 5 mL
UN Number 1993
Formula Weight 99.13
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4,4-Dimethyl-2-oxazoline