missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-2-oxazoline, 98%, Thermo Scientific™
$243.86 - $1,016.66
Chemical Identifiers
| CAS | 30093-99-3 |
|---|---|
| Molecular Formula | C5H9NO |
| Molecular Weight (g/mol) | 99.13 |
| MDL Number | MFCD00014507 |
| InChI Key | KOAMXHRRVFDWRQ-UHFFFAOYSA-N |
| Synonym | 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl |
| PubChem CID | 121630 |
| IUPAC Name | 4,4-dimethyl-4,5-dihydro-1,3-oxazole |
| SMILES | CC1(C)COC=N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC355270100
|
N/A | 10g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC355270500
|
N/A | 50g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 30093-99-3 | |
| 99.13 | |
| KOAMXHRRVFDWRQ-UHFFFAOYSA-N | |
| 121630 | |
| CC1(C)COC=N1 |
| C5H9NO | |
| MFCD00014507 | |
| 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl | |
| 4,4-dimethyl-4,5-dihydro-1,3-oxazole |
Specifications
| 30093-99-3 | |
| 100.0 | |
| 99.0°C to 100.0°C | |
| Authentic | |
| C5H9NO | |
| MFCD00014507 | |
| 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl | |
| CC1(C)COC=N1 | |
| 99.13 | |
| 99.13 | |
| 4, 4-Dimethyl-2-oxazoline |
| 97.5 | |
| 0.9400g/mL | |
| 1°C | |
| 97.5% min. (GC) | |
| 1.4190 to 1.4240 | |
| 0.94 | |
| KOAMXHRRVFDWRQ-UHFFFAOYSA-N | |
| 4,4-dimethyl-4,5-dihydro-1,3-oxazole | |
| 121630 | |
| 98% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
GHS Signal Word: Danger
EINECSNumber : 250-045-7
RUO – Research Use Only