Learn More
4,4-Dimethyl-2-oxazoline, 98%
CAS: 30093-99-3 | C5H9NO | 99.13 g/mol
Supplier: Thermo Scientific Chemicals A1267014
Description
4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.
Solubility
Hydrolyzes with water.
Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
| 4,4-Dimethyl-2-oxazoline | |
| 0.94 | |
| 1°C (33°F) | |
| 1.419 | |
| 25 g | |
| 1209606 | |
| 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl | |
| KOAMXHRRVFDWRQ-UHFFFAOYSA-N | |
| 4,4-dimethyl-5H-1,3-oxazole | |
| 121630 | |
| 98% |
| 30093-99-3 | |
| 99°C to 100°C | |
| C5H9NO | |
| MFCD00014507 | |
| UN1993 | |
| Moisture sensitive | |
| Hydrolyzes with water. | |
| CC1(C)COC=N1 | |
| 99.13 | |
| 99.13 |
Chemical Identifiers
| 30093-99-3 | |
| 99.13 | |
| KOAMXHRRVFDWRQ-UHFFFAOYSA-N | |
| 121630 | |
| CC1(C)COC=N1 |
| C5H9NO | |
| MFCD00014507 | |
| 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl | |
| 4,4-dimethyl-5H-1,3-oxazole |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 250-045-7
RTECSNumber : RP6798385
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only