missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-oxazoline, 98%

Catalog No. AAA1267022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1267022 100 g
AAA1267006 5 g
AAA1267014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1267022 Supplier Thermo Scientific Chemicals Supplier No. A1267022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 30093-99-3 | C5H9NO | 99.13 g/mol

4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 30093-99-3
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00014507
InChI Key KOAMXHRRVFDWRQ-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl
PubChem CID 121630
IUPAC Name 4,4-dimethyl-5H-1,3-oxazole
SMILES CC1(C)COC=N1

Specifications

Density 0.94
Boiling Point 99°C to 100°C
Flash Point 1°C (33°F)
Refractive Index 1.419
Quantity 100 g
UN Number UN1993
Beilstein 1209606
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 99.13
Percent Purity 98%
Chemical Name or Material 4,4-Dimethyl-2-oxazoline
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 250-045-7
RTECSNumber RP6798385
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only