missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethyl-2,2'-bipyridyl 98.0+%, TCI America™
Supplier: TCI America B085225G
Specifications
| 4,4′-Dimethyl-2,2′-bipyridyl | |
| 172°C | |
| MFCD00006441 | |
| 4,4'-dimethyl-2,2'-bipyridine, 4,4'-dimethyl-2,2'-bipyridyl, 4,4'-dimethyl-2,2'-dipyridyl, 2,2'-bi-4-picoline, 2,2'-bipyridine, 4,4'-dimethyl, 2,2'-bi gamma-picoline, 2,2'-di-4-picolyl, 4-methyl-2-4-methylpyridin-2-yl pyridine, 2,2'-bipyridyl, 4,4'-dimethyl, 4,4-dimethyl-2,2-bipyridyl | |
| CC1=CC(=NC=C1)C2=NC=CC(=C2)C | |
| 184.242 | |
| 184.24 | |
| Crystalline Powder |
| 1134-35-6 | |
| C12H12N2 | |
| 25 g | |
| NBPGPQJFYXNFKN-UHFFFAOYSA-N | |
| 4-methyl-2-(4-methylpyridin-2-yl)pyridine | |
| 14338 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 1134-35-6 | |
| 184.242 | |
| NBPGPQJFYXNFKN-UHFFFAOYSA-N | |
| 14338 | |
| CC1=CC(=NC=C1)C2=NC=CC(=C2)C |
| C12H12N2 | |
| MFCD00006441 | |
| 4,4'-dimethyl-2,2'-bipyridine, 4,4'-dimethyl-2,2'-bipyridyl, 4,4'-dimethyl-2,2'-dipyridyl, 2,2'-bi-4-picoline, 2,2'-bipyridine, 4,4'-dimethyl, 2,2'-bi gamma-picoline, 2,2'-di-4-picolyl, 4-methyl-2-4-methylpyridin-2-yl pyridine, 2,2'-bipyridyl, 4,4'-dimethyl, 4,4-dimethyl-2,2-bipyridyl | |
| 4-methyl-2-(4-methylpyridin-2-yl)pyridine |
Safety and Handling
RTECSNumber : DW1765000
TSCA : Yes