missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethyl-2,2'-bipyridyl, 99+%

Catalog No. p-3778380
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC373230050 5 g Glass bottle
AC373230250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC373230050 Supplier Thermo Scientific Chemicals Supplier No. 373230050
Only null left

CAS: 1134-35-6 | C12H12N2 | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1134-35-6
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00006441
InChI Key NBPGPQJFYXNFKN-UHFFFAOYSA-N
Synonym 4,4'-dimethyl-2,2'-bipyridine, 4,4'-dimethyl-2,2'-bipyridyl, 4,4'-dimethyl-2,2'-dipyridyl, 2,2'-bi-4-picoline, 2,2'-bipyridine, 4,4'-dimethyl, 2,2'-bi gamma-picoline, 2,2'-di-4-picolyl, 4-methyl-2-4-methylpyridin-2-yl pyridine, 2,2'-bipyridyl, 4,4'-dimethyl, 4,4-dimethyl-2,2-bipyridyl
PubChem CID 14338
IUPAC Name 4-methyl-2-(4-methylpyridin-2-yl)pyridine
SMILES CC1=CC(=NC=C1)C2=NC=CC(=C2)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 174°C to 176°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 184.24
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Dimethyl-2, 2'-bipyridyl
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-483-2

RUO – Research Use Only