missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-1-pentene, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 426270050

 View more versions of this product

Catalog No. AC426270050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4, 4-Dimethyl-1-pentene
98.5
0.6820g/mL
-12°C
Glass bottle
CH2CHCH2C(CH3)3
5g
4,4-dimethyl-1-pentene, 1-pentene, 4,4-dimethyl, 2,2-dimethyl-4-pentene, 4,4-dimethyl-pent-1-ene, 4,4-dimethyl pent-1-ene, neoheptaene, acmc-1bcwv, ch3 3cch2ch=ch2, 4,4'-dimethyl-1-pentene
CC(C)(C)CC=C
98.19
98.19
762-62-9
100.0
70°C to 72°C
98.5% min. (GC)
C7H14
MFCD00026337
0.682
KLCNJIQZXOQYTE-UHFFFAOYSA-N
4,4-dimethylpent-1-ene
12984
99%

Chemical Identifiers

762-62-9
98.19
KLCNJIQZXOQYTE-UHFFFAOYSA-N
12984
CC(C)(C)CC=C
C7H14
MFCD00026337
4,4-dimethyl-1-pentene, 1-pentene, 4,4-dimethyl, 2,2-dimethyl-4-pentene, 4,4-dimethyl-pent-1-ene, 4,4-dimethyl pent-1-ene, neoheptaene, acmc-1bcwv, ch3 3cch2ch=ch2, 4,4'-dimethyl-1-pentene
4,4-dimethylpent-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-101-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only