missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diisopropylbiphenyl, TCI America™

Catalog No. D204725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D204725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D204725G Supplier TCI America Supplier No. D204725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18970-30-4
Molecular Formula C18H22
Molecular Weight (g/mol) 238.37
MDL Number MFCD00043533
InChI Key NUEUMFZLNOCRCQ-UHFFFAOYSA-N
Synonym 4,4'-diisopropylbiphenyl, 4,4'-diisopropyl-1,1'-biphenyl, 4,4'-di-iso-propylbiphenyl, unii-wr8g413l6a, 1,1'-biphenyl, 4,4'-bis 1-methylethyl, 4,4-diisopropylbiphenyl, 1,1'-biphenyl, 4,4'-bis-1-methylethyl, 1-propan-2-yl-4-4-propan-2-ylphenyl benzene, biphenyl, 4,4'-diisopropyl, pubchem17483
PubChem CID 519615
IUPAC Name 4,4'-bis(propan-2-yl)-1,1'-biphenyl
SMILES CC(C)C1=CC=C(C=C1)C1=CC=C(C=C1)C(C)C

Specifications

Melting Point 66°C
Color White
Quantity 25 g
Formula Weight 238.37
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diisopropylbiphenyl