missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diisopropylbiphenyl, 97%

Catalog No. AAB2077903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2077903 1 g
AAB2077906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2077903 Supplier Thermo Scientific Chemicals Supplier No. B2077903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18970-30-4 | C18H22 | 238.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18970-30-4
Molecular Formula C18H22
Molecular Weight (g/mol) 238.37
MDL Number MFCD00043533
InChI Key NUEUMFZLNOCRCQ-UHFFFAOYSA-N
Synonym 4,4'-diisopropylbiphenyl, 4,4'-diisopropyl-1,1'-biphenyl, 4,4'-di-iso-propylbiphenyl, unii-wr8g413l6a, 1,1'-biphenyl, 4,4'-bis 1-methylethyl, 4,4-diisopropylbiphenyl, 1,1'-biphenyl, 4,4'-bis-1-methylethyl, 1-propan-2-yl-4-4-propan-2-ylphenyl benzene, biphenyl, 4,4'-diisopropyl, pubchem17483
PubChem CID 519615
SMILES CC(C)C1=CC=C(C=C1)C1=CC=C(C=C1)C(C)C

Specifications

Melting Point 66°C
Quantity 1 g
IUPAC Name 1-propan-2-yl-4-(4-propan-2-ylphenyl)benzene
Formula Weight 238.37
Percent Purity 97%
Chemical Name or Material 4,4'-Diisopropylbiphenyl
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only