missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diisocyanato-3,3'-dimethylbiphenyl 98.0+%, TCI America™

Catalog No. p-7127086
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
D205125G 25 g
D2051250G 250 g
2 options

Catalog No. D205125G

Supplier: TCI America D205125G

Only null left

Chemical Identifiers

CAS 91-97-4
Molecular Formula C16H12N2O2
Molecular Weight (g/mol) 264.284
MDL Number MFCD00019908
InChI Key ICLCCFKUSALICQ-UHFFFAOYSA-N
PubChem CID 7072
IUPAC Name 1-isocyanato-4-(4-isocyanato-3-methylphenyl)-2-methylbenzene
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N=C=O)C)N=C=O

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 25 g
Synonym 3,3′C-Dimethylbiphenyl-4,4′C-diyl Diisocyanate
Formula Weight 264.28
Percent Purity ≥98.0% (GC)
Physical Form Pellets
Chemical Name or Material 4,4′-Diisocyanato-3,3′-dimethylbiphenyl
EINECSNumber (4)-0033
RTECSNumber DV3960000
TSCA Yes
Recommended Storage Refrigerator