missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Diiodo-trans-stilbene 97.0+%, TCI America™
$248.34 - $854.77
Chemical Identifiers
| CAS | 201861-91-8 |
|---|---|
| Molecular Formula | C14H10I2 |
| Molecular Weight (g/mol) | 432.043 |
| InChI Key | BYEYAZGFUKIZPM-OWOJBTEDSA-N |
| PubChem CID | 21709796 |
| IUPAC Name | 1-iodo-4-[(E)-2-(4-iodophenyl)ethenyl]benzene |
| SMILES | C1=CC(=CC=C1C=CC2=CC=C(C=C2)I)I |
Chemical Identifiers
| 201861-91-8 | |
| 432.043 | |
| 21709796 | |
| C1=CC(=CC=C1C=CC2=CC=C(C=C2)I)I |
| C14H10I2 | |
| BYEYAZGFUKIZPM-OWOJBTEDSA-N | |
| 1-iodo-4-[(E)-2-(4-iodophenyl)ethenyl]benzene |
Specifications
| 201861-91-8 | |
| 1 g | |
| C1=CC(=CC=C1C=CC2=CC=C(C=C2)I)I | |
| 432.043 | |
| 432.04 | |
| Crystalline Powder |
| C14H10I2 | |
| BYEYAZGFUKIZPM-OWOJBTEDSA-N | |
| 1-iodo-4-[(E)-2-(4-iodophenyl)ethenyl]benzene | |
| 21709796 | |
| ≥97.0% (GC) | |
| 4,4′-Diiodo-trans-stilbene |
Safety and Handling
TSCA : No